Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075271.1 | NP_001068739.1 | 2 - 3 | 570 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Bos taurus Probe spans exons |