Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLA2G16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075280.2 | NP_001068748.1 | 2 - 3 | 118 | 94 | |
GenBank mRNA | BC118454.1 | - | 2 - 3 | 119 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |