Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARPC1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075359.1 | NP_001068827.1 | 7 - 8 | 904 | 81 | |
GenBank mRNA | BC147946.1 | - | 7 - 8 | 973 | 81 | |
BT025475.1 | - | 7 - 8 | 904 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |