Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP3S2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075376.1 | NP_001068844.1 | 3 - 4 | 310 | 129 | |
GenBank mRNA | BC116115.1 | - | 3 - 4 | 310 | 129 | |
BT025371.1 | - | 3 - 4 | 286 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |