Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOMM40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075438.1 | NP_001068906.1 | 6 - 7 | 703 | 81 | |
GenBank mRNA | BC116105.1 | - | 6 - 7 | 703 | 81 | |
HM347497.1 | - | 5 - 6 | 647 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |