Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASSF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075442.1 | NP_001068910.1 | 2 - 3 | 323 | 84 | |
GenBank mRNA | BC116028.1 | - | 2 - 3 | 323 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |