Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BID | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075446.2 | NP_001068914.1 | 3 - 4 | 428 | 110 | |
GenBank mRNA | AB238940.1 | - | 1 - 2 | 213 | 110 | |
BC118359.1 | - | 3 - 4 | 430 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |