Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075569.1 | NP_001069037.1 | 1 - 2 | 419 | 167 | |
GenBank mRNA | BC118302.1 | - | 1 - 2 | 419 | 167 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |