Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075594.1 | NP_001069062.1 | 2 - 3 | 329 | 83 | |
GenBank mRNA | BC118261.1 | - | 2 - 3 | 329 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |