Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQCD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075606.1 | NP_001069074.1 | 2 - 3 | 914 | 60 | |
GenBank mRNA | BC118347.1 | - | 2 - 3 | 914 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |