Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNIP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075612.2 | NP_001069080.1 | 3 - 4 | 377 | 100 | |
GenBank mRNA | BC118417.1 | - | 3 - 4 | 378 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |