Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APOO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075639.1 | NP_001069107.1 | 4 - 5 | 317 | 143 | |
GenBank mRNA | BC118339.1 | - | 4 - 5 | 317 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |