Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM39B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075658.1 | NP_001069126.1 | 4 - 5 | 493 | 71 | |
GenBank mRNA | BC118147.1 | - | 4 - 5 | 493 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |