Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBOX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075669.1 | NP_001069137.1 | 1 - 2 | 69 | 85 | |
GenBank mRNA | BC114651.1 | - | 1 - 2 | 69 | 85 |
Gene Symbol: FASTKD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206420.1 | NP_001193349.1 | 1 - 2 | 69 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |