Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YJU2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075691.1 | NP_001069159.1 | 7 - 8 | 909 | 62 | |
GenBank mRNA | BC118290.1 | - | 7 - 8 | 909 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |