Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UQCC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075714.2 | NP_001069182.1 | 1 - 2 | 139 | 58 | |
GenBank mRNA | BC118350.1 | - | 1 - 2 | 140 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |