Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFRP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075764.1 | NP_001069232.1 | 3 - 4 | 849 | 71 | |
GenBank mRNA | BC118245.1 | - | 3 - 4 | 849 | 71 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |