Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABPB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075804.2 | NP_001069272.1 | 8 - 9 | 1155 | 86 | |
GenBank mRNA | BC114881.1 | - | 8 - 9 | 1156 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |