Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075829.1 | NP_001069297.1 | 5 - 6 | 603 | 68 | |
GenBank mRNA | BC116110.1 | - | 5 - 6 | 603 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |