Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCXR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075891.1 | NP_001069359.1 | 1 - 2 | 105 | 94 | |
GenBank mRNA | BC142210.1 | - | 1 - 2 | 61 | 94 | |
BT025479.1 | - | 1 - 2 | 105 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |