Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BEGAIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075902.2 | NP_001069370.2 | 5 - 6 | 384 | 88 | |
GenBank mRNA | BC114844.1 | - | 5 - 6 | 385 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |