Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LY9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075918.1 | NP_001069386.1 | 6 - 7 | 1251 | 83 | |
GenBank mRNA | BC118207.1 | - | 6 - 7 | 1251 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |