Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC116108.1 | - | 6 - 7 | 954 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |