Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076013.2 | NP_001069481.1 | 3 - 4 | 302 | 134 | |
GenBank mRNA | BC116151.1 | - | 3 - 4 | 303 | 134 | |
GQ367549.1 | - | 3 - 4 | 291 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |