Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076013.2 | NP_001069481.1 | 8 - 9 | 686 | 82 | |
GenBank mRNA | BC116151.1 | - | 8 - 9 | 687 | 82 | |
GQ367549.1 | - | 8 - 9 | 675 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Inventoried Probe spans exons |