Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP2S1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076078.2 | NP_001069546.1 | 1 - 2 | 70 | 92 | |
GenBank mRNA | BC118437.1 | - | 1 - 2 | 71 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |