Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P4HTM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076115.2 | NP_001069583.1 | 3 - 4 | 617 | 86 | |
GenBank mRNA | BC114843.1 | - | 3 - 4 | 618 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |