Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076119.1 | NP_001069587.1 | 1 - 2 | 447 | 93 | |
GenBank mRNA | BC118327.1 | - | 1 - 2 | 447 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |