Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COQ10A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076130.2 | NP_001069598.1 | 1 - 2 | 294 | 127 | |
GenBank mRNA | BC114817.1 | - | 1 - 2 | 295 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |