Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GDAP1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076261.1 | NP_001069729.1 | 4 - 5 | 730 | 73 | |
GenBank mRNA | BC133446.1 | - | 4 - 5 | 660 | 73 | |
BT025400.1 | - | 4 - 5 | 730 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |