Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GDAP1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076261.1 | NP_001069729.1 | 6 - 7 | 1528 | 146 | |
GenBank mRNA | BT025400.1 | - | 6 - 7 | 1528 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Bos taurus Probe spans exons |