Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076262.1 | NP_001069730.1 | 2 - 3 | 642 | 124 | |
GenBank mRNA | BC142425.1 | - | 2 - 3 | 629 | 124 | |
BC151822.1 | - | 2 - 3 | 629 | 124 | ||
BT025450.1 | - | 2 - 3 | 642 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |