Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076291.1 | NP_001069759.1 | 5 - 6 | 631 | 130 | |
GenBank mRNA | BC118262.1 | - | 5 - 6 | 631 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |