Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLNS1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076300.1 | NP_001069768.1 | 4 - 5 | 565 | 147 | |
GenBank mRNA | BC116139.1 | - | 4 - 5 | 565 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |