Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2G2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076328.1 | NP_001069796.1 | 1 - 2 | 94 | 71 | |
GenBank mRNA | BC118376.1 | - | 1 - 2 | 94 | 71 | |
BT026143.1 | - | 1 - 2 | 73 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |