Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIP4P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076359.1 | NP_001069827.1 | 6 - 7 | 750 | 63 | |
GenBank mRNA | BC118333.1 | - | 6 - 7 | 750 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |