Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076480.2 | NP_001069948.1 | 2 - 3 | 668 | 83 | |
GenBank mRNA | BC114805.1 | - | 2 - 3 | 669 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |