Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANGPT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076483.2 | NP_001069951.1 | 7 - 8 | 1751 | 81 | |
GenBank mRNA | BC114033.1 | - | 7 - 8 | 1753 | 81 | |
EU599227.1 | - | 7 - 8 | 1491 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |