Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076821.1 | NP_001070289.1 | 1 - 2 | 437 | 121 | |
GenBank mRNA | BC113310.1 | - | 1 - 2 | 437 | 121 | |
BT021038.1 | - | 2 - 3 | 280 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |