Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARMCX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076960.1 | NP_001070428.1 | 2 - 3 | 159 | 77 | |
GenBank mRNA | BC114723.1 | - | 2 - 3 | 159 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |