Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP4A11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077908.1 | NP_001071376.1 | 2 - 3 | 376 | 81 | |
GenBank mRNA | BC118405.1 | - | 2 - 3 | 376 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |