Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUBA4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078158.1 | NP_001071626.1 | 2 - 3 | 278 | 95 | |
GenBank mRNA | BC118199.1 | - | 2 - 3 | 278 | 95 | |
BT021034.1 | - | 2 - 3 | 260 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |