Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TM2D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079593.2 | NP_001073061.1 | 1 - 2 | 177 | 147 | |
GenBank mRNA | BC112745.1 | - | 1 - 2 | 178 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |