Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBLN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079603.1 | NP_001073071.1 | 2 - 3 | 525 | 70 | |
GenBank mRNA | BC118385.1 | - | 2 - 3 | 525 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |