Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VIM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173969.3 | NP_776394.2 | 7 - 8 | 1400 | 76 | |
GenBank mRNA | BC118269.1 | - | 7 - 8 | 1400 | 76 | |
L13263.1 | - | 7 - 8 | 1272 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |