Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FTH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174062.4 | NP_776487.1 | 1 | 608 | 82 | |
GenBank mRNA | AB003093.1 | - | 3 - 4 | 548 | 82 | |
AB098768.1 | - | 1 - 2 | 317 | 82 | ||
AB098915.1 | - | 3 - 4 | 410 | 82 | ||
AF540563.1 | - | 3 - 4 | 356 | 82 | ||
AY911329.1 | - | 3 - 4 | 608 | 82 | ||
BC105376.1 | - | 1 | 609 | 82 | ||
BC151549.1 | - | 3 - 4 | 610 | 82 | ||
JX051225.1 | - | 1 | 156 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Both primers and probe map within a single exon |