Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLAMF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174184.4 | NP_776609.2 | 5 - 6 | 950 | 124 | |
GenBank mRNA | AF329970.1 | - | 5 - 6 | 889 | 124 | |
BC114833.1 | - | 5 - 6 | 951 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |