Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IMPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174361.4 | NP_776786.1 | 2 - 3 | 119 | 83 | |
GenBank mRNA | BC118411.1 | - | 2 - 3 | 120 | 83 | |
BT030590.2 | - | 2 - 3 | 85 | 83 | ||
J05394.1 | - | 2 - 3 | 129 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |