Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IMPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174361.4 | NP_776786.1 | 3 - 4 | 253 | 113 | |
GenBank mRNA | BC118411.1 | - | 3 - 4 | 254 | 113 | |
BT030590.2 | - | 3 - 4 | 219 | 113 | ||
J05394.1 | - | 3 - 4 | 263 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |