Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKAG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174586.2 | NP_777011.2 | 10 - 11 | 889 | 83 | |
GenBank mRNA | BC113296.1 | - | 10 - 11 | 889 | 83 | |
BT025456.1 | - | 10 - 11 | 892 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |